Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCGGGCATCAAACACTAGCCAG[A/G]GGTCCATGTCAGCTGCCATGGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602289 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf68 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf68 - chromosome 11 open reading frame 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135635.1 | 259 | Missense Mutation | CCC,CTC | P,L 78 | NP_001129107.1 | |
NM_031450.3 | 259 | Missense Mutation | CCC,CTC | P,L 77 | NP_113638.2 |
DRAP1 - DR1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |