Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCCCTATTGAGGAGGGGCCAGT[C/T]CCCAGAACAGGGGCAAAAGGGCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLOD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLOD5 - glyoxalase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080489.2 | 534 | Silent Mutation | GTC,GTT | V,V 130 | NP_001073958.2 | |
XM_011543911.2 | 534 | Silent Mutation | GTC,GTT | V,V 88 | XP_011542213.1 | |
XM_011543912.2 | 534 | Silent Mutation | GTC,GTT | V,V 153 | XP_011542214.1 |