Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCAACTACACGCTGCGCTACGC[C/G]CGCCCTGCGGACGCGCTCTGAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYRM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYRM4 - LYR motif containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164840.2 | 1056 | Intron | NP_001158312.1 | |||
NM_001164841.2 | 1056 | Intron | NP_001158313.1 | |||
NM_001318782.1 | 1056 | Intron | NP_001305711.1 | |||
NM_001318783.1 | 1056 | Intron | NP_001305712.1 | |||
NM_020408.5 | 1056 | Intron | NP_065141.3 | |||
XM_017011083.1 | 1056 | Intron | XP_016866572.1 | |||
XM_017011084.1 | 1056 | Intron | XP_016866573.1 |
LYRM4-AS1 - LYRM4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R3G - protein phosphatase 1 regulatory subunit 3G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145115.2 | 1056 | Silent Mutation | GCC,GCG | A,A 352 | NP_001138587.1 |