Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCATCGGAAGGCTCGAGTCCTCTC[A/G]CGGACAAGCTGAACGAACACATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614139 MIM: 608998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAPPC12 - trafficking protein particle complex 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321102.1 | 811 | Missense Mutation | ACG,GCG | T,A 59 | NP_001308031.1 | |
NM_016030.5 | 811 | Missense Mutation | ACG,GCG | T,A 59 | NP_057114.5 | |
XM_011510350.2 | 811 | Missense Mutation | ACG,GCG | T,A 59 | XP_011508652.1 | |
XM_011510352.2 | 811 | Missense Mutation | ACG,GCG | T,A 59 | XP_011508654.1 | |
XM_011510353.2 | 811 | Missense Mutation | ACG,GCG | T,A 59 | XP_011508655.1 | |
XM_011510354.2 | 811 | Missense Mutation | ACG,GCG | T,A 59 | XP_011508656.1 | |
XM_011510355.2 | 811 | Intron | XP_011508657.1 | |||
XM_011510356.1 | 811 | Intron | XP_011508658.1 | |||
XM_017004257.1 | 811 | Missense Mutation | ACG,GCG | T,A 59 | XP_016859746.1 |
TSSC1 - tumor suppressing subtransferable candidate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |