Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCCTAAATGATAATCAGTGCC[C/T]CAGCAAGTGGTGTCATTGAGGTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603712 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM20 - ADAM metallopeptidase domain 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003814.4 | 11067 | Nonsense Mutation | TGA,TGG | *,W 642 | NP_003805.3 | |
XM_005268151.3 | 11067 | Nonsense Mutation | TGA,TGG | *,W 642 | XP_005268208.1 |