Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCCGTCTCTATACTCAGGGTCA[C/T]CCTGCCACTGAGGTTTGCTTCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615329 MIM: 609713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOC2 - exocyst complex component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018303.5 | 585 | Intron | NP_060773.3 | |||
XM_017011018.1 | 585 | Intron | XP_016866507.1 | |||
XM_017011019.1 | 585 | Intron | XP_016866508.1 | |||
XM_017011020.1 | 585 | Intron | XP_016866509.1 | |||
XM_017011021.1 | 585 | Intron | XP_016866510.1 | |||
XM_017011022.1 | 585 | Intron | XP_016866511.1 | |||
XM_017011023.1 | 585 | Intron | XP_016866512.1 | |||
XM_017011024.1 | 585 | Intron | XP_016866513.1 | |||
XM_017011025.1 | 585 | Intron | XP_016866514.1 | |||
XM_017011026.1 | 585 | Intron | XP_016866515.1 |
HUS1B - HUS1 checkpoint clamp component B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_148959.3 | 585 | Missense Mutation | ATG,GTG | M,V 189 | NP_683762.2 |