Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGTTGAGTCGCTTGAGGATCTG[C/G]GCCAACACGTTCACTGTCTGCTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613305 MIM: 189968 MIM: 602921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALKBH7 - alkB homolog 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GTF2F1 - general transcription factor IIF subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002096.2 | 1767 | Silent Mutation | GCC,GCG | A,A 494 | NP_002087.2 |
MIR6885 - microRNA 6885 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSPN - persephin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |