Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGCCCCAGAGGACCTCGAGGCG[C/T]GTGTGGTTCCGAGAACCCCCAGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf180 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf180 - chromosome 14 open reading frame 180 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008404.2 | 540 | Silent Mutation | CGC,CGT | R,R 68 | NP_001008404.1 | |
NM_001286399.1 | 540 | Silent Mutation | CGC,CGT | R,R 68 | NP_001273328.1 | |
NM_001286400.1 | 540 | Silent Mutation | CGC,CGT | R,R 68 | NP_001273329.1 | |
XM_005267638.3 | 540 | Silent Mutation | CGC,CGT | R,R 68 | XP_005267695.1 | |
XM_011536764.2 | 540 | Silent Mutation | CGC,CGT | R,R 68 | XP_011535066.1 |
TMEM179 - transmembrane protein 179 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |