Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTCCAGGTGTCTGGATGGCCC[C/G]GGGTAGCTTCTGGCTGGTCAGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNPPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNPPD1 - cyclin Pas1/PHO80 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM134A - family with sequence similarity 134 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NHEJ1 - non-homologous end joining factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC23A3 - solute carrier family 23 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144889.1 | 505 | Missense Mutation | CGG,GGG | R,G 132 | NP_001138361.1 | |
NM_001144890.1 | 505 | Missense Mutation | CGG,GGG | R,G 132 | NP_001138362.1 | |
NM_144712.4 | 505 | Missense Mutation | CGG,GGG | R,G 132 | NP_653313.3 |