Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGCCAGAGAGTGAGGGAGAGGC[A/G]AAAGAAGGAAAGTCAGAGAGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEAL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCEAL4 - transcription elongation factor A like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006935.2 | 507 | Silent Mutation | GCA,GCG | A,A 71 | NP_001006936.1 | |
NM_001006937.2 | 507 | Silent Mutation | GCA,GCG | A,A 71 | NP_001006938.1 | |
NM_001300901.1 | 507 | Silent Mutation | GCA,GCG | A,A 214 | NP_001287830.1 | |
NM_001305840.1 | 507 | Silent Mutation | GCA,GCG | A,A 71 | NP_001292769.1 | |
NM_001305841.1 | 507 | Silent Mutation | GCA,GCG | A,A 71 | NP_001292770.1 | |
NM_001305842.1 | 507 | Silent Mutation | GCA,GCG | A,A 71 | NP_001292771.1 | |
NM_024863.5 | 507 | Silent Mutation | GCA,GCG | A,A 71 | NP_079139.4 | |
XM_017029856.1 | 507 | Silent Mutation | GCA,GCG | A,A 133 | XP_016885345.1 | |
XM_017029857.1 | 507 | Silent Mutation | GCA,GCG | A,A 71 | XP_016885346.1 |