Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATAATCAGCTCTTAGTTGCTTTA[A/C]GCTGACAGGAGAAGGGATGCTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613369 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC47 - coiled-coil domain containing 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX42 - DEAD-box helicase 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007372.3 | Intron | NP_031398.2 | ||||
NM_203499.2 | Intron | NP_987095.1 | ||||
XM_006721657.1 | Intron | XP_006721720.1 | ||||
XM_011524259.2 | Intron | XP_011522561.1 | ||||
XM_017024111.1 | Intron | XP_016879600.1 | ||||
XM_017024112.1 | Intron | XP_016879601.1 |