Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCGCGGCCACCCGGAGCCTTCC[A/G]TGTCCTGGAGGAAGGACGGTGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608630 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ROBO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ROBO3 - roundabout guidance receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022370.3 | 761 | Missense Mutation | ATG,GTG | M,V 198 | NP_071765.2 | |
XM_017018122.1 | 761 | Missense Mutation | ATG,GTG | M,V 176 | XP_016873611.1 |