Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAGGTAAATGAGGGCCAGCAAG[C/T]GCCTGTCCATGCGGTGAGGGTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLPH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOLPH3 - golgi phosphoprotein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022130.3 | 1258 | Missense Mutation | CAC,CGC | H,R 235 | NP_071413.1 | |
XM_017009686.1 | 1258 | Missense Mutation | CAC,CGC | H,R 143 | XP_016865175.1 | |
XM_017009687.1 | 1258 | Missense Mutation | CAC,CGC | H,R 143 | XP_016865176.1 |