Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGACCCCTTACTTAGACCTCAA[C/G]CTGGAGACAGGGGTGCTGTACGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608286 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927359 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927359 - uncharacterized LOC101927359 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCDH10 - protocadherin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020815.2 | 1052 | Missense Mutation | AAC,AAG | N,K 67 | NP_065866.1 | |
NM_032961.2 | 1052 | Missense Mutation | AAC,AAG | N,K 67 | NP_116586.1 | |
XM_011532150.1 | 1052 | Missense Mutation | AAC,AAG | N,K 67 | XP_011530452.1 |