Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACTGTCAGGGAGGGATAGGAAA[C/T]GAGAGTAGCCTCAGTGCTGCTGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600911 | ||||||||||||||||||||
Literature Links: |
MPPED2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPPED2 - metallophosphoesterase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145399.1 | 2663 | UTR 3 | NP_001138871.1 | |||
NM_001584.2 | 2663 | Intron | NP_001575.1 | |||
XM_005253110.3 | 2663 | Intron | XP_005253167.1 | |||
XM_005253111.2 | 2663 | Intron | XP_005253168.1 | |||
XM_005253112.1 | 2663 | Intron | XP_005253169.1 | |||
XM_005253114.3 | 2663 | Intron | XP_005253171.1 | |||
XM_011520346.1 | 2663 | Intron | XP_011518648.1 | |||
XM_011520347.2 | 2663 | Intron | XP_011518649.1 | |||
XM_017018231.1 | 2663 | Intron | XP_016873720.1 | |||
XM_017018232.1 | 2663 | UTR 3 | XP_016873721.1 | |||
XM_017018233.1 | 2663 | Intron | XP_016873722.1 | |||
XM_017018234.1 | 2663 | Intron | XP_016873723.1 | |||
XM_017018235.1 | 2663 | Intron | XP_016873724.1 | |||
XM_017018236.1 | 2663 | Intron | XP_016873725.1 | |||
XM_017018237.1 | 2663 | Intron | XP_016873726.1 |