Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGTTTCTCGACAACATTGTAACT[A/G]CATTTCTTCAAGTGGTCTGCCATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609101 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO30 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO30 - F-box protein 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032145.4 | 7179 | Silent Mutation | TGC,TGT | C,C 707 | NP_115521.3 | |
XM_005267159.3 | 7179 | Silent Mutation | TGC,TGT | C,C 707 | XP_005267216.1 | |
XM_011536177.2 | 7179 | Silent Mutation | TGC,TGT | C,C 707 | XP_011534479.1 | |
XM_017011353.1 | 7179 | Silent Mutation | TGC,TGT | C,C 707 | XP_016866842.1 |
LOC100507557 - uncharacterized LOC100507557 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |