Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATACTTCCTAAAGAAACTTCAT[C/T]GTCATCCAGGTTACCAAGTTCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612828 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEBPZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEBPZ - CCAAT/enhancer binding protein zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005760.2 | 2725 | Missense Mutation | NP_005751.2 |
CEBPZOS - CEBPZ opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322373.1 | 2725 | Intron | NP_001309302.1 | |||
NM_001322374.1 | 2725 | Intron | NP_001309303.1 | |||
NM_001322375.1 | 2725 | Intron | NP_001309304.1 | |||
NM_001322376.1 | 2725 | Intron | NP_001309305.1 | |||
XM_017003106.1 | 2725 | Intron | XP_016858595.1 | |||
XM_017003107.1 | 2725 | Intron | XP_016858596.1 | |||
XM_017003108.1 | 2725 | Intron | XP_016858597.1 | |||
XM_017003109.1 | 2725 | Intron | XP_016858598.1 |