Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTTAGGAGTATTTTATCAATCA[A/G]CTGAAGATAGAAGGAGTTGAACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC4 - ERCC excision repair 4, endonuclease catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005236.2 | 200 | Silent Mutation | CAA,CAG | Q,Q 75 | NP_005227.1 | |
XM_011522424.2 | 200 | Silent Mutation | CAA,CAG | Q,Q 75 | XP_011520726.1 | |
XM_011522427.1 | 200 | Intron | XP_011520729.1 | |||
XM_017023043.1 | 200 | UTR 5 | XP_016878532.1 |