Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATGGTGCACACCTCATCGATG[A/T]GCACAGCGCGCTCCGTACGTGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607204 | ||||||||||||||||||||
Literature Links: |
PUM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PUM1 - pumilio RNA binding family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001020658.1 | 3409 | Missense Mutation | CAC,CTC | H,L 1099 | NP_001018494.1 | |
NM_014676.2 | 3409 | Missense Mutation | CAC,CTC | H,L 1097 | NP_055491.1 |
SNORD103A - small nucleolar RNA, C/D box 103A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |