Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGTGGGGGGCGGCGGCACTGC[C/G]GGCGGGGACCGCTGGGAAGGCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTDSPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTDSPL2 - CTD small phosphatase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF3J - eukaryotic translation initiation factor 3 subunit J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284335.1 | 229 | Silent Mutation | GCC,GCG | A,A 34 | NP_001271264.1 | |
NM_001284336.1 | 229 | Silent Mutation | GCC,GCG | A,A 34 | NP_001271265.1 | |
NM_003758.3 | 229 | Silent Mutation | GCC,GCG | A,A 34 | NP_003749.2 |
EIF3J-AS1 - EIF3J antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |