Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCTTCCCCAGGGACAAGGCTC[A/G]ACCCTTCACAGTTCCCATCACTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BNC1 - basonuclin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301206.1 | 2914 | Missense Mutation | TCG,TTG | S,L 890 | NP_001288135.1 | |
NM_001717.3 | 2914 | Missense Mutation | TCG,TTG | S,L 897 | NP_001708.3 | |
XM_011521893.1 | 2914 | Missense Mutation | TCG,TTG | S,L 872 | XP_011520195.1 | |
XM_011521894.1 | 2914 | Missense Mutation | TCG,TTG | S,L 779 | XP_011520196.1 |