Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACACACAGCCTTGCGCACCTCCT[C/T]ACCAAATACGCTGAGCAGCTGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605847 MIM: 600435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL7C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL7C - BCL tumor suppressor 7C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CTF1 - cardiotrophin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142544.1 | 257 | Silent Mutation | CTC,CTT | L,L 36 | NP_001136016.1 | |
NM_001330.3 | 257 | Silent Mutation | CTC,CTT | L,L 37 | NP_001321.1 | |
XM_011545759.2 | 257 | Silent Mutation | CTC,CTT | L,L 59 | XP_011544061.1 | |
XM_011545760.2 | 257 | Silent Mutation | CTC,CTT | L,L 45 | XP_011544062.1 |
MIR762 - microRNA 762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR762HG - MIR762 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |