Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCCCCCATCCCTTCTCCCCCTA[C/T]CCTATCTCTCTCTTTCCACCTGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607206 MIM: 608059 MIM: 610407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOXE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALOXE3 - arachidonate lipoxygenase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HES7 - hes family bHLH transcription factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165967.1 | 2788 | UTR 3 | NP_001159439.1 | |||
NM_032580.3 | 2788 | UTR 3 | NP_115969.2 | |||
XM_011524041.2 | 2788 | UTR 3 | XP_011522343.1 | |||
XM_017025232.1 | 2788 | UTR 3 | XP_016880721.1 |
TRG-GCC2-6 - transfer RNA-Gly (GCC) 2-6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |