Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGATCAATGAGATGGTGAACAGG[A/G]CCAAACGGAAAGCAGGGGTGGATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606828 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAGK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAGK - N-acetylglucosamine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017567.4 | 537 | Missense Mutation | ACC,GCC | T,A 102 | NP_060037.3 | |
XM_005264412.4 | 537 | Missense Mutation | ACC,GCC | T,A 5 | XP_005264469.1 | |
XM_005264413.1 | 537 | Missense Mutation | ACC,GCC | T,A 5 | XP_005264470.1 | |
XM_005264415.2 | 537 | Missense Mutation | ACC,GCC | T,A 5 | XP_005264472.1 | |
XM_011532957.1 | 537 | Missense Mutation | ACC,GCC | T,A 5 | XP_011531259.1 | |
XM_017004440.1 | 537 | Missense Mutation | ACC,GCC | T,A 102 | XP_016859929.1 |