Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGCTTTACCGTTGTGAGTTGT[C/T]TGGGTAATCGTGCCTGGTTTTAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604477 MIM: 600124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBX3 - chromobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007276.4 | Intron | NP_009207.2 | ||||
NM_016587.3 | Intron | NP_057671.2 | ||||
XM_005249611.3 | Intron | XP_005249668.1 |
HNRNPA2B1 - heterogeneous nuclear ribonucleoprotein A2/B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |