Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGCCCCCATCAGCACCGGCCTT[C/T]CCTGTCTCTGGCTGTTCCAGGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3H4 - zinc finger CCCH-type containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015168.1 | 4919 | Silent Mutation | GGA,GGG | G,G 1202 | NP_055983.1 | |
XM_005258676.3 | 4919 | Silent Mutation | GGA,GGG | G,G 1166 | XP_005258733.1 | |
XM_005258677.4 | 4919 | Silent Mutation | GGA,GGG | G,G 1161 | XP_005258734.1 | |
XM_005258678.2 | 4919 | Silent Mutation | GGA,GGG | G,G 1153 | XP_005258735.1 | |
XM_006723113.3 | 4919 | Silent Mutation | GGA,GGG | G,G 1202 | XP_006723176.1 | |
XM_011526668.2 | 4919 | Silent Mutation | GGA,GGG | G,G 1161 | XP_011524970.1 | |
XM_011526669.2 | 4919 | Silent Mutation | GGA,GGG | G,G 1156 | XP_011524971.1 | |
XM_011526670.2 | 4919 | Silent Mutation | GGA,GGG | G,G 1152 | XP_011524972.1 | |
XM_017026529.1 | 4919 | Silent Mutation | GGA,GGG | G,G 1189 | XP_016882018.1 | |
XM_017026530.1 | 4919 | Silent Mutation | GGA,GGG | G,G 1118 | XP_016882019.1 | |
XM_017026531.1 | 4919 | Silent Mutation | GGA,GGG | G,G 905 | XP_016882020.1 |