Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTGGTCACCACCAGCACGCTGC[A/G]GAACTTGTCAAACAGCATGAGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156565 MIM: 600928 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPG - N-methylpurine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015052.2 | 1966 | Intron | NP_001015052.1 | |||
NM_001015054.2 | 1966 | Intron | NP_001015054.1 | |||
NM_002434.3 | 1966 | Intron | NP_002425.2 |
NPRL3 - NPR3 like, GATOR1 complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039476.2 | 1966 | Missense Mutation | CGC,TGC | R,C 369 | NP_001034565.1 | |
NM_001077350.2 | 1966 | Missense Mutation | CGC,TGC | R,C 548 | NP_001070818.1 | |
NM_001243247.1 | 1966 | Missense Mutation | CGC,TGC | R,C 470 | NP_001230176.1 | |
NM_001243248.1 | 1966 | Missense Mutation | CGC,TGC | R,C 523 | NP_001230177.1 | |
NM_001243249.1 | 1966 | Missense Mutation | CGC,TGC | R,C 523 | NP_001230178.1 |