Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGTTTGTTCATATGCCATACCT[C/T]TTTCCTTAGCTTTTTGATGGTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYAR - Ly1 antibody reactive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145725.1 | 1177 | Missense Mutation | AAG,GAG | K,E 335 | NP_001139197.1 | |
NM_017816.2 | 1177 | Missense Mutation | AAG,GAG | K,E 335 | NP_060286.1 | |
XM_011513505.1 | 1177 | Missense Mutation | AAG,GAG | K,E 335 | XP_011511807.1 | |
XM_011513506.2 | 1177 | Missense Mutation | AAG,GAG | K,E 335 | XP_011511808.1 |