Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACCACTAAGGCAGAGAGGGACT[C/G]TGGGAAGCTCTCCATCGAGTCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605772 MIM: 611568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EBAG9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EBAG9 - estrogen receptor binding site associated, antigen, 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYBU - syntabulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099743.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 533 | NP_001093213.1 | |
NM_001099744.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 534 | NP_001093214.1 | |
NM_001099745.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 534 | NP_001093215.1 | |
NM_001099746.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 415 | NP_001093216.1 | |
NM_001099747.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 533 | NP_001093217.1 | |
NM_001099748.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 534 | NP_001093218.1 | |
NM_001099749.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 415 | NP_001093219.1 | |
NM_001099750.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 534 | NP_001093220.1 | |
NM_001099751.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 533 | NP_001093221.1 | |
NM_001099752.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 534 | NP_001093222.1 | |
NM_001099753.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 533 | NP_001093223.1 | |
NM_001099754.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 534 | NP_001093224.1 | |
NM_001099755.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 415 | NP_001093225.1 | |
NM_001099756.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 531 | NP_001093226.1 | |
NM_017786.5 | 1870 | Missense Mutation | CAG,GAG | Q,E 533 | NP_060256.3 | |
XM_005250999.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 404 | XP_005251056.1 | |
XM_005251000.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 371 | XP_005251057.1 | |
XM_011517154.2 | 1870 | Missense Mutation | CAG,GAG | Q,E 367 | XP_011515456.1 | |
XM_017013614.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 415 | XP_016869103.1 | |
XM_017013615.1 | 1870 | Missense Mutation | CAG,GAG | Q,E 415 | XP_016869104.1 |