Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGATGGGTACGACGGACTGCC[A/G]GGGCCCAAGGGGGAGCCAGGTGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120550 MIM: 120570 MIM: 120575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QA - complement C1q A chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1QB - complement C1q B chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1QC - complement C1q C chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114101.1 | 288 | Silent Mutation | CCA,CCG | P,P 54 | NP_001107573.1 | |
NM_172369.3 | 288 | Silent Mutation | CCA,CCG | P,P 54 | NP_758957.2 |