Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCTCACCACAAGGACCACCCCT[A/G]CAGTGGTGCCCACCAGGGGTGGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603060 MIM: 607883 | ||||||||||||||||||||
Literature Links: |
KIF1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF1C - kinesin family member 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC52A1 - solute carrier family 52 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104577.1 | 1831 | Missense Mutation | GCA,GTA | A,V 373 | NP_001098047.1 | |
NM_017986.3 | 1831 | Missense Mutation | GCA,GTA | A,V 373 | NP_060456.3 | |
XM_011523951.1 | 1831 | Missense Mutation | GCA,GTA | A,V 373 | XP_011522253.1 |