Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCGCGGCGGGCGCAGGGCGGCC[C/G]GAGCCCTGGGAGCTGTCCCTGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609217 | ||||||||||||||||||||
Literature Links: |
LOC107984817 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107984817 - uncharacterized LOC107984817 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIRE2 - spire type actin nucleation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032451.1 | 109 | Silent Mutation | CCC,CCG | P,P 17 | NP_115827.1 | |
XM_011523379.1 | 109 | Intron | XP_011521681.1 | |||
XM_011523380.2 | 109 | UTR 5 | XP_011521682.1 | |||
XM_011523381.1 | 109 | Intron | XP_011521683.1 | |||
XM_011523384.1 | 109 | Silent Mutation | CCC,CCG | P,P 17 | XP_011521686.1 | |
XM_011523385.1 | 109 | Silent Mutation | CCC,CCG | P,P 17 | XP_011521687.1 | |
XM_017023779.1 | 109 | Intron | XP_016879268.1 | |||
XM_017023780.1 | 109 | Intron | XP_016879269.1 | |||
XM_017023781.1 | 109 | Silent Mutation | CCC,CCG | P,P 17 | XP_016879270.1 |