Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGGAAGAAGGGGCTGCGGATGT[C/G]AGGATCCAGGCCAGGGGGTGTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA21 - spermatogenesis associated 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198546.1 | 2304 | Missense Mutation | CAC,GAC | H,D 437 | NP_940948.1 | |
XM_011541407.2 | 2304 | Missense Mutation | CAC,GAC | H,D 663 | XP_011539709.1 | |
XM_011541417.2 | 2304 | Intron | XP_011539719.1 | |||
XM_017001216.1 | 2304 | Missense Mutation | CAC,GAC | H,D 663 | XP_016856705.1 | |
XM_017001217.1 | 2304 | Missense Mutation | CAC,GAC | H,D 619 | XP_016856706.1 | |
XM_017001218.1 | 2304 | Intron | XP_016856707.1 | |||
XM_017001219.1 | 2304 | Intron | XP_016856708.1 | |||
XM_017001220.1 | 2304 | Missense Mutation | CAC,GAC | H,D 487 | XP_016856709.1 | |
XM_017001221.1 | 2304 | Missense Mutation | CAC,GAC | H,D 414 | XP_016856710.1 | |
XM_017001222.1 | 2304 | Missense Mutation | CAC,GAC | H,D 414 | XP_016856711.1 | |
XM_017001223.1 | 2304 | Missense Mutation | CAC,GAC | H,D 414 | XP_016856712.1 | |
XM_017001224.1 | 2304 | Missense Mutation | CAC,GAC | H,D 414 | XP_016856713.1 | |
XM_017001225.1 | 2304 | Missense Mutation | CAC,GAC | H,D 352 | XP_016856714.1 |
SZRD1 - SUZ RNA binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |