Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGCCTCCTCAGCATTGAGTGTC[C/T]GTGTCTTCCTGAGATTCTTCAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TARSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TARSL2 - threonyl-tRNA synthetase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152334.2 | 1886 | Missense Mutation | CAG,CGG | Q,R 794 | NP_689547.2 | |
XM_006720386.3 | 1886 | UTR 3 | XP_006720449.1 | |||
XM_017021912.1 | 1886 | Intron | XP_016877401.1 | |||
XM_017021913.1 | 1886 | Missense Mutation | CAG,CGG | Q,R 551 | XP_016877402.1 |
TM2D3 - TM2 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |