Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGATCCAAGCTATTGTCCTGCCC[A/G]TGGCTTCCCATCTCAGGACCCTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116896 MIM: 605300 | ||||||||||||||||||||
Literature Links: |
CUX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUX1 - cut like homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4285 - microRNA 4285 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2B2 - SH2B adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020979.4 | 123 | Missense Mutation | CAT,CGT | H,R 9 | NP_066189.3 | |
XM_005276975.4 | 123 | Missense Mutation | CAT,CGT | H,R 9 | XP_005277032.1 | |
XM_005276976.3 | 123 | Missense Mutation | CAT,CGT | H,R 9 | XP_005277033.1 | |
XM_005276977.4 | 123 | Intron | XP_005277034.1 | |||
XM_005276979.3 | 123 | Intron | XP_005277036.1 | |||
XM_011515740.2 | 123 | Intron | XP_011514042.1 | |||
XM_017011680.1 | 123 | Intron | XP_016867169.1 | |||
XM_017011681.1 | 123 | Intron | XP_016867170.1 |