Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCCTAATTCCTAGCTACATTC[A/G]TGATTCAACTATTGCAGTGGTTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 301770 MIM: 300521 MIM: 300632 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARR3 - arrestin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIF4A - kinesin family member 4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDZD11 - PDZ domain containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB41 - RAB41, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032726.2 | 1545 | Missense Mutation | CAT,CGT | H,R 101 | NP_001027898.2 | |
XM_011530948.2 | 1545 | Missense Mutation | CAT,CGT | H,R 102 | XP_011529250.1 | |
XM_017029488.1 | 1545 | Missense Mutation | CAT,CGT | H,R 102 | XP_016884977.1 |