Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGGAAAGCAGTACCTTTTAAA[A/T]CTCATTGATACACCGGTAAGTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUF1 - GUF1 homolog, GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021927.2 | 472 | Missense Mutation | AAA,AAT | K,N 137 | NP_068746.2 | |
XM_005248122.3 | 472 | UTR 5 | XP_005248179.1 | |||
XM_011513732.2 | 472 | Missense Mutation | AAA,AAT | K,N 137 | XP_011512034.1 |