Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAATAGGAGGAAAAATGACCAC[A/G]TTGAAGGTGAGTAGAGCTTGCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100379224 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100379224 - uncharacterized LOC100379224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF224 - zinc finger protein 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF225 - zinc finger protein 225 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321685.1 | 450 | Silent Mutation | ACA,ACG | T,T 3 | NP_001308614.1 | |
NM_013362.3 | 450 | Silent Mutation | ACA,ACG | T,T 3 | NP_037494.2 | |
XM_005259223.3 | 450 | Intron | XP_005259280.1 | |||
XM_011527285.2 | 450 | Silent Mutation | ACA,ACG | T,T 3 | XP_011525587.1 | |
XM_011527286.2 | 450 | Silent Mutation | ACA,ACG | T,T 3 | XP_011525588.1 |