Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MX1 - MX dynamin like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144925.2 | Intron | NP_001138397.1 | ||||
NM_001178046.2 | Intron | NP_001171517.1 | ||||
NM_001282920.1 | Intron | NP_001269849.1 | ||||
NM_002462.4 | Intron | NP_002453.2 | ||||
XM_005260978.4 | Intron | XP_005261035.1 | ||||
XM_005260979.2 | Intron | XP_005261036.1 | ||||
XM_005260980.2 | Intron | XP_005261037.1 | ||||
XM_005260981.2 | Intron | XP_005261038.1 | ||||
XM_005260982.2 | Intron | XP_005261039.1 | ||||
XM_011529568.2 | Intron | XP_011527870.1 | ||||
XM_011529570.2 | Intron | XP_011527872.1 | ||||
XM_017028349.1 | Intron | XP_016883838.1 | ||||
XM_017028350.1 | Intron | XP_016883839.1 | ||||
XM_017028351.1 | Intron | XP_016883840.1 | ||||
XM_017028352.1 | Intron | XP_016883841.1 |