Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTAGATAAGGTTACTAGAAGA[C/T]AGGGAATCCTGGGCTTATCTGGATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603028 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100419170 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100419170 - toll like receptor 2 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLR2 - toll like receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318787.1 | 308 | Intron | NP_001305716.1 | |||
NM_001318789.1 | 308 | Intron | NP_001305718.1 | |||
NM_001318790.1 | 308 | Intron | NP_001305719.1 | |||
NM_001318791.1 | 308 | Intron | NP_001305720.1 | |||
NM_001318793.1 | 308 | Intron | NP_001305722.1 | |||
NM_001318795.1 | 308 | Intron | NP_001305724.1 | |||
NM_001318796.1 | 308 | Intron | NP_001305725.1 | |||
NM_003264.4 | 308 | Intron | NP_003255.2 | |||
XM_011532215.2 | 308 | Intron | XP_011530517.1 | |||
XM_011532216.2 | 308 | Intron | XP_011530518.1 | |||
XM_017008573.1 | 308 | Intron | XP_016864062.1 | |||
XM_017008574.1 | 308 | Intron | XP_016864063.1 | |||
XM_017008575.1 | 308 | UTR 5 | XP_016864064.1 | |||
XM_017008576.1 | 308 | Intron | XP_016864065.1 |