Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191191 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFRSF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFRSF1B - TNF receptor superfamily member 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001066.2 | Intron | NP_001057.1 | ||||
XM_011542060.2 | Intron | XP_011540362.1 | ||||
XM_011542063.2 | Intron | XP_011540365.1 | ||||
XM_017002211.1 | Intron | XP_016857700.1 | ||||
XM_017002212.1 | Intron | XP_016857701.1 | ||||
XM_017002213.1 | Intron | XP_016857702.1 | ||||
XM_017002214.1 | Intron | XP_016857703.1 | ||||
XM_017002215.1 | Intron | XP_016857704.1 |