Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM164 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM164 - transmembrane protein 164 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017698.2 | Intron | NP_060168.2 | ||||
NM_032227.3 | Intron | NP_115603.2 | ||||
XM_005262205.4 | Intron | XP_005262262.1 | ||||
XM_005262206.3 | Intron | XP_005262263.1 | ||||
XM_005262208.4 | Intron | XP_005262265.1 | ||||
XM_011531051.2 | Intron | XP_011529353.1 | ||||
XM_011531054.1 | Intron | XP_011529356.1 | ||||
XM_011531055.2 | Intron | XP_011529357.1 | ||||
XM_017029894.1 | Intron | XP_016885383.1 | ||||
XM_017029895.1 | Intron | XP_016885384.1 | ||||
XM_017029896.1 | Intron | XP_016885385.1 | ||||
XM_017029897.1 | Intron | XP_016885386.1 | ||||
XM_017029898.1 | Intron | XP_016885387.1 | ||||
XM_017029899.1 | Intron | XP_016885388.1 | ||||
XM_017029900.1 | Intron | XP_016885389.1 |