Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NHSL2 - NHS like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013627.2 | Intron | NP_001013649.2 | ||||
XM_011530932.1 | Intron | XP_011529234.1 | ||||
XM_011530933.1 | Intron | XP_011529235.1 | ||||
XM_011530934.2 | Intron | XP_011529236.1 | ||||
XM_017029473.1 | Intron | XP_016884962.1 | ||||
XM_017029474.1 | Intron | XP_016884963.1 | ||||
XM_017029475.1 | Intron | XP_016884964.1 | ||||
XM_017029476.1 | Intron | XP_016884965.1 | ||||
XM_017029477.1 | Intron | XP_016884966.1 |