Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300286 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLF8 - Kruppel like factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159296.2 | Intron | NP_001152768.1 | ||||
NM_001324099.1 | Intron | NP_001311028.1 | ||||
NM_001324100.1 | Intron | NP_001311029.1 | ||||
NM_001324102.1 | Intron | NP_001311031.1 | ||||
NM_001324104.1 | Intron | NP_001311033.1 | ||||
NM_001324105.1 | Intron | NP_001311034.1 | ||||
NM_007250.5 | Intron | NP_009181.2 | ||||
XM_005261977.2 | Intron | XP_005262034.1 | ||||
XM_005261979.3 | Intron | XP_005262036.1 | ||||
XM_006724575.2 | Intron | XP_006724638.1 | ||||
XM_006724576.3 | Intron | XP_006724639.1 | ||||
XM_011530759.2 | Intron | XP_011529061.1 | ||||
XM_017029250.1 | Intron | XP_016884739.1 |
Set Membership: |
HapMap |