Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD21L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD21L1 - RAD21 cohesin complex component like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136566.2 | Intron | NP_001130038.2 | ||||
XM_006723602.2 | Intron | XP_006723665.1 | ||||
XM_006723603.2 | Intron | XP_006723666.1 | ||||
XM_006723604.2 | Intron | XP_006723667.1 | ||||
XM_006723605.3 | Intron | XP_006723668.1 | ||||
XM_011529317.2 | Intron | XP_011527619.1 | ||||
XM_011529319.1 | Intron | XP_011527621.1 | ||||
XM_011529320.2 | Intron | XP_011527622.1 |