Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603343 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTRNR2L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTRNR2L3 - MT-RNR2-like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAE1 - ribonucleic acid export 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015885.1 | Intron | NP_001015885.1 | ||||
NM_003610.3 | Intron | NP_003601.1 | ||||
XM_005260582.2 | Intron | XP_005260639.2 | ||||
XM_005260583.2 | Intron | XP_005260640.1 | ||||
XM_011529087.2 | Intron | XP_011527389.1 | ||||
XM_011529088.2 | Intron | XP_011527390.1 | ||||
XM_011529089.1 | Intron | XP_011527391.1 | ||||
XM_017028108.1 | Intron | XP_016883597.1 |