Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602498 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TFG - TRK-fused gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007565.2 | Intron | NP_001007566.1 | ||||
NM_001195478.1 | Intron | NP_001182407.1 | ||||
NM_001195479.1 | Intron | NP_001182408.1 | ||||
NM_006070.5 | Intron | NP_006061.2 | ||||
XM_005247066.1 | Intron | XP_005247123.1 | ||||
XM_006713472.1 | Intron | XP_006713535.1 | ||||
XM_006713473.1 | Intron | XP_006713536.1 | ||||
XM_011512334.1 | Intron | XP_011510636.1 | ||||
XM_017005527.1 | Intron | XP_016861016.1 | ||||
XM_017005528.1 | Intron | XP_016861017.1 | ||||
XM_017005529.1 | Intron | XP_016861018.1 | ||||
XM_017005530.1 | Intron | XP_016861019.1 |
Set Membership: |
HapMap |