Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605290 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OPA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OPA1 - OPA1, mitochondrial dynamin like GTPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015560.2 | Intron | NP_056375.2 | ||||
NM_130831.2 | Intron | NP_570844.1 | ||||
NM_130832.2 | Intron | NP_570845.1 | ||||
NM_130833.2 | Intron | NP_570846.1 | ||||
NM_130834.2 | Intron | NP_570847.2 | ||||
NM_130835.2 | Intron | NP_570848.1 | ||||
NM_130836.2 | Intron | NP_570849.2 | ||||
NM_130837.2 | Intron | NP_570850.2 | ||||
XM_017006500.1 | Intron | XP_016861989.1 |
Set Membership: |
HapMap |