Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605489 | |||||||||||||||||||||||
Literature Links: |
IFT81 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
IFT81 - intraflagellar transport 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143779.1 | Intron | NP_001137251.1 | ||||
NM_014055.3 | Intron | NP_054774.2 | ||||
NM_031473.3 | Intron | NP_113661.2 | ||||
XM_011538232.2 | Intron | XP_011536534.1 | ||||
XM_017019217.1 | Intron | XP_016874706.1 | ||||
XM_017019218.1 | Intron | XP_016874707.1 |