Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604010 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELMSAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELMSAN1 - ELM2 and Myb/SANT domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001043318.2 | 5354 | UTR 3 | NP_001036783.1 | |||
NM_194278.3 | 5354 | UTR 3 | NP_919254.2 | |||
XM_005268204.1 | 5354 | UTR 3 | XP_005268261.1 | |||
XM_005268205.1 | 5354 | UTR 3 | XP_005268262.1 | |||
XM_005268206.1 | 5354 | UTR 3 | XP_005268263.1 |
PNMA1 - paraneoplastic Ma antigen 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |